Anti-GPRC5C Rabbit Polyclonal Antibody

48-243
10075-836EA 684.46 USD
10075-836
Anti-GPRC5C Rabbit Polyclonal Antibody
Antibodies
GPRC5C is an Orphan-C GPCR induced by retinoids.

GPRC5C antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPRC5C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR