Anti-GPR174 Rabbit Polyclonal Antibody

48-242
10075-834EA 674.53 USD
10075-834
Anti-GPR174 Rabbit Polyclonal Antibody
Antibodies
GPR174/JEG18 is an Orphan-A GPCR with an unknown ligand.

GPR174 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR174
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR