Anti-PRLHR Rabbit Polyclonal Antibody

48-229
10077-912EA 674.53 USD
10077-912
Anti-PRLHR Rabbit Polyclonal Antibody
Antibodies
Prolactin releasing hormone receptor (hGR3/GPR10) is a Releasing Hormone Receptor that controls the secretion of prolactin from the anterior pituitary.

PRLHR antibody can be used in Western Blot, immunohistochemistry starting at 20 ?g/mL, and flow cytometry.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PRLHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR