Anti-GPR132 Rabbit Polyclonal Antibody

Supplier: Prosci
48-227
10077-908EA 785.68 USD
10077-908
Anti-GPR132 Rabbit Polyclonal Antibody
Antibodies
G2A belongs to the Lysophospholipid/Lysosphingolipid Receptor family and is an anti-proliferative cell cycle regulator that plays a critical role in controlling peripheral lymphocyte homeostasis. Loss of G2A expression leads to the development of a late-onset autoimmune syndrome (Le et al, 2001). Lysophosphatidylcholine (LPC) is the activating ligand for G2A.

GPR132 antibody can be used in ELISA, and immunohistochemistry starting at 28 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR132
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR