Anti-TMEM5 Rabbit Polyclonal Antibody

48-202
10077-862EA 674.53 USD
10077-862
Anti-TMEM5 Rabbit Polyclonal Antibody
Antibodies
TMEM5 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TMEM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR