Anti-PTPMT1 Rabbit Polyclonal Antibody

48-193
10077-844EA 698.16 USD
10077-844
Anti-PTPMT1 Rabbit Polyclonal Antibody
Antibodies
Provisional protein phosphatase.

PTPMT1 antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunoprecipitation.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PTPMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Dog, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR