Anti-PYGB Rabbit Polyclonal Antibody

Supplier: Prosci
48-190
10074-318EA 785.68 USD
10074-318
Anti-PYGB Rabbit Polyclonal Antibody
Antibodies
PYGB antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, and flow cytometry starting at 1:10.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PYGB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Chicken, Drosophila, Xenopus, Gorilla, Hamster, Human, Monkey, Mouse, Orangutan, Pig, Rabbit, Rat, Sheep, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR