Anti-CAV1 Rabbit Polyclonal Antibody

48-184
10074-306EA 698.16 USD
10074-306
Anti-CAV1 Rabbit Polyclonal Antibody
Antibodies
Caveolin 1 is the main component of the caveolae plasma membranes found in most cell types. Caveolin 1 functions as a membrane adaptor to link integrin subunits to the tyrosine kinase FYN, an initiating step in coupling integrins to the Ras-ERK pathway and promoting cell cycle progression. CAV1 may also be a tumor suppressor gene and a negative regulator of the Ras-p42/44 MAP kinase cascade.

Caveolin-1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, immunofluorescence starting at 10 ?g/mL, and immunoprecipitation.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CAV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Chimpanzee, Dog, Guinea pig, Gorilla, Horse, Human, Monkey, Mouse, Orangutan, Pig, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR