Anti-PTPRM Rabbit Polyclonal Antibody

48-180
10074-298EA 674.53 USD
10074-298
Anti-PTPRM Rabbit Polyclonal Antibody
Antibodies
PTP mu, a R2A receptor protein tyrosine phosphatase, is composed of an extracellular segment containing a MAM domain, an immunoglobulin domain and four fibronectin type III repeats, a transmembrane segment, and two intracellular protein phosphatase domains. PTP mu has been shown to affect cell-cell aggregation, regulate cardiac myocyte Kv1.5 channel expression, and promote neurite outgrowth of retinal ganglion cells. The human PTP mu gene is localized on chromosome 18pter-q11, a region with frequent abnormalities implicated in human cancer. PTP mu may be important in responses to pathological conditions associated with the loss of cell-cell interactions in the heart.

PTPRM antibody can be used in immunohistochemistry starting at 5 ?g/mL, and flow cytometry.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PTPRM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Dog, Gorilla, Human, Monkey, Orangutan, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR