Anti-EPHA4 Rabbit Polyclonal Antibody

48-176
10074-290EA 698.16 USD
10074-290
Anti-EPHA4 Rabbit Polyclonal Antibody
Antibodies
EPHA4 antibody can be used in Western Blot, immunohistochemistry starting at 1:200, and flow cytometry starting at 1:100 - 1:50.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EPHA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR