Anti-EPHA4 Rabbit Polyclonal Antibody

Supplier: Prosci
48-175
10074-288EA 785.68 USD
10074-288
Anti-EPHA4 Rabbit Polyclonal Antibody
Antibodies
Nak1, a NR4 Nerve Growth Factor-Like receptor, has been shown to affect the hypothalamic-pituitary axis, neuronal differentiation, neuroendocrine regulation of adrenocortical function, and T-cell apoptosis. Nak1 also is known as nuclear receptor Tr3 and is similar to mouse Nur77 and rat NGFI-B. It is encoded by one of the immediate early genes (IEG) in response to nerve growth factor and other serum growth factors. Nak1 can bind to DNA as a monomer, homodimer, or heterodimer (with retinoid X receptors). Nak1 has been shown to inhibit vascular lesion formation and could serve as a novel target in the treatment of vascular diseases.

EPHA4 antibody can be used in immunohistochemistry starting at 7.3 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EPHA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR