Nak1, a NR4 Nerve Growth Factor-Like receptor, has been shown to affect the hypothalamic-pituitary axis, neuronal differentiation, neuroendocrine regulation of adrenocortical function, and T-cell apoptosis. Nak1 also is known as nuclear receptor Tr3 and is similar to mouse Nur77 and rat NGFI-B. It is encoded by one of the immediate early genes (IEG) in response to nerve growth factor and other serum growth factors. Nak1 can bind to DNA as a monomer, homodimer, or heterodimer (with retinoid X receptors). Nak1 has been shown to inhibit vascular lesion formation and could serve as a novel target in the treatment of vascular diseases.
NR4A1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: NR4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Horse, Human, Monkey, Orangutan