Anti-SLC5A3 Rabbit Polyclonal Antibody

48-167
10077-840EA 674.35 USD
10077-840
Anti-SLC5A3 Rabbit Polyclonal Antibody
Antibodies
SLC5A3 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC5A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR