Anti-GPR115 Rabbit Polyclonal Antibody

48-165
10077-836EA 540.97 USD
10077-836
Anti-GPR115 Rabbit Polyclonal Antibody
G-protein coupled receptor GPR115 is an Orphan-B GPCR with an unknown ligand.

GPR115 antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR115
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR