Anti-BRS3 Rabbit Polyclonal Antibody

48-161
10077-828EA 684.46 USD
10077-828
Anti-BRS3 Rabbit Polyclonal Antibody
Antibodies
BRS3 is classified as an Orphan-A Receptor and is highly similar to the bombesin receptors but has a very low affinity for bombesin itself. Mice lacking BRS3 develop metabolic defects and obesity, suggesting that BRS3 may be an important target for obesity research. In addition, BRS3 may be involved in diabetes and hypertension.

BRS3 antibody can be used in immunohistochemistry starting at 1:50, immunofluorescence, and flow cytometry.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BRS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR