Anti-PYGL Rabbit Polyclonal Antibody

48-157
10077-820EA 674.53 USD
10077-820
Anti-PYGL Rabbit Polyclonal Antibody
Antibodies
PYGL antibody can be used in Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PYGL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR