Anti-GPR108 Rabbit Polyclonal Antibody

48-150
10077-806EA 684.46 USD
10077-806
Anti-GPR108 Rabbit Polyclonal Antibody
Antibodies
LUSTR2 is an Orphan-U GPCR with an unknown ligand.

GPR108 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR108
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR