Anti-HTR5A Rabbit Polyclonal Antibody

48-149
10077-804EA 698.16 USD
10077-804
Anti-HTR5A Rabbit Polyclonal Antibody
Antibodies
The 5-HT5A Receptor is a member of the Serotonin Receptor subfamily.

HTR5A antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HTR5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Dog, Guinea pig, Gorilla, Horse, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR