Anti-GPR146 Rabbit Polyclonal Antibody

48-142
10077-790EA 698.16 USD
10077-790
Anti-GPR146 Rabbit Polyclonal Antibody
Antibodies
GPR146 is an Orphan-U GPCR with an unknown ligand, but is likely to be a chemokine receptor.

GPR146 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR146
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR