Anti-NPBWR1 Rabbit Polyclonal Antibody

48-136
10077-778EA 684.46 USD
10077-778
Anti-NPBWR1 Rabbit Polyclonal Antibody
Antibodies
GPR7 is a Neuropeptide Receptor GPCR. GPR7 is implicated in the regulation of feeding, neuroendorine system, memory, and learning.

NPBWR1 antibody can be used in immunohistochemistry starting at 17 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NPBWR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR