Anti-GPR61 Rabbit Polyclonal Antibody

48-135
10077-776EA 684.46 USD
10077-776
Anti-GPR61 Rabbit Polyclonal Antibody
Antibodies
GPR61 is an Orphan-A GPCR with an unknown ligand.

GPR61 antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR