Anti-GPR75 Rabbit Polyclonal Antibody

Supplier: Prosci

48-131
10077-768EA 582.9 USD
10077-768
Anti-GPR75 Rabbit Polyclonal Antibody
Antibodies
GPR75 is an Orphan-A GPCR with an unknown ligand.

GPR75 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR