Anti-XCR1 Rabbit Polyclonal Antibody

Supplier: Prosci
48-130
10077-766EA 785.68 USD
10077-766
Anti-XCR1 Rabbit Polyclonal Antibody
Antibodies
CCXCR1 is a Chemokine Receptor expressed in lymphotactin-responsive leukocyte cells. The viral macrophage inflammatory protein-II, vMIP-II (but not vMIP-I), has been found to be an antagonist of lymphotactin activity at CCXCR1. The CCXCR1 gene is located near the genes for C-C chemokine receptor 9 and SIV/HIV receptor Bonzo on chromosome 3. CCXCR1 is associated with inflammatory and immunological responses by regulating the trafficking of T and B lymphocytes and neutrophils. The up-regulation of CCXCR1 in peripheral T cells is correlated with the inflammatory process observed in Crohns disease.

XCR1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: XCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR