Anti-MCHR1 Rabbit Polyclonal Antibody

Supplier: Prosci
48-128
10077-762EA 785.68 USD
10077-762
Anti-MCHR1 Rabbit Polyclonal Antibody
Antibodies
MCHR1 is a Melanin-Concentrating Hormone Receptor that binds MCH to regulate nutritional homeostasis, including feeding and body adiposity. The receptor also functions in sensory perception and integration of smell and gustatory inputs, as well as in emotion and memory.

MCHR1 antibody can be used in ELISA, and immunohistochemistry starting at 15 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MCHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Chimpanzee, Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR