Anti-PTGDR2 Rabbit Polyclonal Antibody

Supplier: Prosci
48-122
10077-750EA 785.68 USD
10077-750
Anti-PTGDR2 Rabbit Polyclonal Antibody
Antibodies
GPR44, also called CRTH2, is a Prostanoid Receptor that binds prostaglandin D2. PGD2 signals through CRTH2 to induce Th2, eosinophil, and basophil chemotaxis in a G-alpha(i) dependent manner. CRTH2 may play a role in mast cell-mediated immune responses such as those involved in allergic reactions. This receptor is upregulated in activated T helper-2 (Th2) cells, peripheral blood basophils, and eosinophils, but not in Th1 lineage cells. It may be a useful marker for Th2 and Tc2 cells.

GPR44 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PTGDR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR