Anti-TPRA1 Rabbit Polyclonal Antibody

48-121
10077-748EA 684.46 USD
10077-748
Anti-TPRA1 Rabbit Polyclonal Antibody
Antibodies
GPR175 is an Orphan-U GPCR with an unknown ligand. LOC51210 has been implicated in the effects of ischemic hypoxia and reoxygenation.

GPR175 antibody can be used in immunohistochemistry starting at 10 ?g/mL, immunoprecipitation, and flow cytometry.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TPRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Dog, Gorilla, Hamster, Horse, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR