Anti-CHRM3 Rabbit Polyclonal Antibody

48-117
10077-740EA 698.16 USD
10077-740
Anti-CHRM3 Rabbit Polyclonal Antibody
Antibodies
Muscarinic acetylcholine receptor 3 is an Acetylcholine Receptor that controls smooth muscle contraction by mediating phosphoinositide hydrolysis and Ca2+ mobilization. Activation of this receptor causes secretion in glandular tissues. Mouse studies suggest that the M3 receptor may play a role in appetite stimulation and food intake.

CHRM3 antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CHRM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR