Anti-STEAP1 Mouse Monoclonal Antibody [clone: [J2D2]]

48-100
10074-260EA 684.46 USD
10074-260
Anti-STEAP1 Mouse Monoclonal Antibody [clone: [J2D2]]
Antibodies
STEAP1 antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STEAP1
Clonality: Monoclonal
Clone: [J2D2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR