Anti-NAPA Mouse Monoclonal Antibody

Supplier: Prosci

48-097
10074-254EA 582.9 USD
10074-254
Anti-NAPA Mouse Monoclonal Antibody
Antibodies
SNAP antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2, 0.1 mM PMSF, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Napa
Clonality: Monoclonal
Clone: [6B7-3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Bovine, Dog, Hamster, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR