Anti-CFL1 Mouse Monoclonal Antibody [clone: [AT1C1]]

48-090
10074-240EA 674.53 USD
10074-240
Anti-CFL1 Mouse Monoclonal Antibody [clone: [AT1C1]]
Antibodies
Cofilin antibody can be used in ELISA, Western Blot starting at 0.5 - 2 ?g/mL, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation starting at 3 - 5 ng/1E6 cells

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CFL1
Clonality: Monoclonal
Clone: [AT1C1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR