Anti-APP Mouse Monoclonal Antibody [clone: [J4H9]]

48-089
10074-238EA 684.46 USD
10074-238
Anti-APP Mouse Monoclonal Antibody [clone: [J4H9]]
Antibodies
APP antibody can be used in Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: [J4H9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR