Anti-YWHAQ Mouse Monoclonal Antibody [clone: [AT1A1]]

Supplier: Prosci

48-087
10074-234EA 582.74 USD
10074-234
Anti-YWHAQ Mouse Monoclonal Antibody [clone: [AT1A1]]
Antibodies
14-3-3 Tau antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: YWHAQ
Clonality: Monoclonal
Clone: [AT1A1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR