Anti-HSF1 Mouse Monoclonal Antibody [clone: [J7F9]]

Supplier: Prosci

48-071
10077-730EA 582.9 USD
10077-730
Anti-HSF1 Mouse Monoclonal Antibody [clone: [J7F9]]
Antibodies
HSF1 antibody can be used in Western Blot starting at 0.5 - 2 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSF1
Clonality: Monoclonal
Clone: [J7F9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR