Anti-YWHAE Mouse Monoclonal Antibody

48-068
10075-796EA 491.23 USD
10075-796
Anti-YWHAE Mouse Monoclonal Antibody
14-3-3 Epsilon antibody can be used in Western Blot starting at 2.5 - 5 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: YWHAE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR