Anti-IRF7 Mouse Monoclonal Antibody [clone: [3D9]]

Supplier: Prosci
48-065
10075-792EA 785.68 USD
10075-792
Anti-IRF7 Mouse Monoclonal Antibody [clone: [3D9]]
Antibodies
IRF7 antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IRF7
Clonality: Monoclonal
Clone: [3D9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR