Anti-VAMP2 Mouse Monoclonal Antibody [clone: [3E5]]

48-056
10075-774EA 674.35 USD
10075-774
Anti-VAMP2 Mouse Monoclonal Antibody [clone: [3E5]]
Antibodies
VAMP2 antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: VAMP2
Clonality: Monoclonal
Clone: [3E5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR