Anti-HSPB1 Mouse Monoclonal Antibody [clone: [2A5]]

48-052
10075-766EA 684.46 USD
10075-766
Anti-HSPB1 Mouse Monoclonal Antibody [clone: [2A5]]
Antibodies
HSPB1 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSPB1
Clonality: Monoclonal
Clone: [2A5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR