Anti-NPM Mouse Monoclonal Antibody [clone: [5E3]]

48-050
10077-722EA 674.53 USD
10077-722
Anti-NPM Mouse Monoclonal Antibody [clone: [5E3]]
Antibodies
NPM1 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NPM1
Clonality: Monoclonal
Clone: [5E3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR