Anti-ENO2 Mouse Monoclonal Antibody [clone: [NSE-P1]]

48-035
10077-692EA 540.97 USD
10077-692
Anti-ENO2 Mouse Monoclonal Antibody [clone: [NSE-P1]]
NSE antibody can be used in immunohistochemistry starting at 5 - 10 ?g/mL.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ENO2
Clonality: Monoclonal
Clone: [NSE-P1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR