Anti-CFH Mouse Monoclonal Antibody

Supplier: Prosci

48-034
10077-690EA 561.89 USD
10077-690
Anti-CFH Mouse Monoclonal Antibody
Antibodies
Factor H antibody can be used in immunohistochemistry starting at 2.5 - 5 ?g/mL.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CFH
Clonality: Monoclonal
Clone: [OX-24]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR