Anti-ITGB1 Mouse Monoclonal Antibody

48-023
10077-670EA 674.53 USD
10077-670
Anti-ITGB1 Mouse Monoclonal Antibody
Antibodies
Integrin Beta 1 antibody can be used in Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 10 ?g/mL, immunocytochemistry, and flow cytometry.

PBS, pH 7.4, 0.09% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR