Anti-IL1R1 Mouse Monoclonal Antibody [clone: [40101]]

48-022
10077-668EA 698.16 USD
10077-668
Anti-IL1R1 Mouse Monoclonal Antibody [clone: [40101]]
Antibodies
IL1R1 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.4

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: [40101]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR