Anti-PTPRC Mouse Monoclonal Antibody

48-008
10075-748EA 674.53 USD
10075-748
Anti-PTPRC Mouse Monoclonal Antibody
Antibodies
CD45RA antibody can be used in Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation.

PBS, sodium azide, a stabilizing agent.

Add glycerol (40-50%) and store at -20 °C. For short term store at 4 °C.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR