Anti-RB1 Rabbit Polyclonal Antibody

Supplier: Prosci
47-180
10069-732EA 701.96 USD
10069-732
Anti-RB1 Rabbit Polyclonal Antibody
Antibodies
Epithelium-specific gene regulation plays acritical role during embryogenesis, and the epithelial cell lineage is the first differentiated cell type to appear afterfertilization. Until recently, very few epithelial cell-restricted transcription factors had been described. Members of the ets transcription factor/oncogene family havebeen implicated in human carcinogenesis following the discovery of several distinctchromosomal translocations involving members of the ets family in different types of cancer. ets factors play key roles intranscriptional control of stringently regulated genes, such as genes involved intissue development, differentiation,angiogenesis, cell cycle control and cellproliferation. ESE-1 is the first member of the ets family that was found to be exclusively expressed and inducible inepithelial cells.

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR