Anti-RB1 Mouse Monoclonal Antibody [clone: 1320]

Supplier: Prosci

47-179
10070-994EA 615.28 USD
10070-994
Anti-RB1 Mouse Monoclonal Antibody [clone: 1320]
Antibodies
Protein G-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RB1
Clonality: Monoclonal
Clone: 1320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR