Anti-RB1 Rabbit Polyclonal Antibody

47-177
10071-760EA 640.19 USD
10071-760
Anti-RB1 Rabbit Polyclonal Antibody
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR