Anti-SHPRH Rabbit Polyclonal Antibody

47-170
10069-730EA 625.45 USD
10069-730
Anti-SHPRH Rabbit Polyclonal Antibody
Antibodies
Affinity-purified

PBS containing 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Shprh
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR