Anti-MUC12 Mouse Monoclonal Antibody [clone: 10H5]

Supplier: Prosci

47-153
10070-988EA 665.42 USD
10070-988
Anti-MUC12 Mouse Monoclonal Antibody [clone: 10H5]
Antibodies
Protein G-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MUC12
Clonality: Monoclonal
Clone: 10H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1/kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR