Anti-CDKL3 Mouse Monoclonal Antibody [clone: 12F3.3]

Supplier: Prosci

47-152
10071-724EA 615.28 USD
10071-724
Anti-CDKL3 Mouse Monoclonal Antibody [clone: 12F3.3]
Antibodies
Protein G-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CDKL3
Clonality: Monoclonal
Clone: 12F3.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a/kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR