Anti-KLK11 Rabbit Polyclonal Antibody

Supplier: Prosci
47-139
10071-006EA 701.96 USD
10071-006
Anti-KLK11 Rabbit Polyclonal Antibody
Antibodies
NKX3.1 is a homeobox gene which exhibits prostate- and testis-specific expression. Loss of NKX3.1 expression has been implicated in prostate development and tumorigenesis. NKX3.1 expression is dramatically down-regulated in testicular cancer of germ cell origin. Results suggest that NKX3.1 is essential for normal testis function and that its loss of expression is highly associated with the invasive phenotype of testicular germ cell tumors.

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KLK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR